Learn More
TARGETMOL CHEMICALS INC Xevinapant 10mg
SDP
Supplier:  TARGETMOL CHEMICALS INC T676310MG
Encompass_Preferred
AT406 (SM-406, ARRY-334543) is a potent Smac mimetic and an antagonist of IAP (inhibitor of apoptosis protein via E3 ubiquitin ligase), binding to XIAP-BIR3, cIAP1-BIR3 and cIAP2-BIR3 with Ki of 66.4 nM, 1.9 nM, and 5.1 nM, 50- to 100-fold higher affinities than the Smac AVPI peptide. Phase 1.
Catalog No. NC1665743