Learn More
Medchemexpress LLC AT6; PROTAC AT1 analogue | 2098836-50-9 | 99.9% | 1004.68 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1123751MG
Encompass_Preferred
AT6 is a PROTAC AT1 analogue, a PROTAC connected by ligands for von Hippel-Lindau and BRD4, demonstrating high selectivity for bromodomain (Brd4). PROTACs are bifunctional molecules that recruit a target protein in proximity to an E3 ubiquitin ligase to trigger protein degradation. This compound's ligand folds into itself, enabling specific intermolecular interactions in the ternary complex.
  • Exhibits high selectivity to bromodomain (Brd4)
  • Bifunctional molecules that recruit a target protein in proximity to an E3 ubiquitin ligase
  • Triggers protein degradation
  • Ligand folds to allow specific intermolecular interactions
  • Shows selective depletion of Brd4 in cells
Catalog No. 50-004-38378