Learn More
Selleck Chemical LLC AT7867 10mg 857531-00-1
SDP
Supplier:  Selleck Chemical LLC S155810MG
Encompass_Preferred
AT7867 is a potent ATP-competitive inhibitor of Akt1/2/3 and p70S6K/PKA with IC50 of 32 nM/17 nM/47 nM and 85 nM/20 nM in cell-free assays, respectively; little activity outside the AGC kinase family. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-882-4