Learn More
Medchemexpress LLC HY-124603 5mg , AT791 CAS:1219962-49-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1246035MG
Encompass_Preferred
HY-124603 5mg AT791 CAS: 1219962-49-8 AT791 is a potent and orally bioavailable TLR7 and TLR9 inhibitor. AT791 inhibits TLR7 and 9 signaling in a variety of human and mouse cell types and inhibits DNA-TLR9 interaction in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9998