Learn More
TARGETMOL CHEMICALS INC AT9283 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T306810MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AT9283 (J-504568) is an effective multi-targeted inhibitor of JAK2(IC50=1.2 nM) and JAK3(IC50=1.1 nM) Aurora A Aurora B and Abl(T315I). purity: 99%
Catalog No. 50-258-0397