Learn More
TARGETMOL CHEMICALS INC Atabecestat 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T143381MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Atabecestat (JNJ-54861911) is a potent, brain-penetrant, and orally active beta-site amyloid precursor protein cleaving enzyme 1 (BACE1) inhibitor that achieves robust and high CSF Abeta reduction, is well-tolerated, and displays sustained pharmacokinetic (PK) and pharmacodynamic (PD) characteristics. Atabecestat (JNJ-54861911) has the potential for Alzheimer's Disease treatment[1]. Purity 99.77%
Catalog No. 50-288-0124