Learn More
Selleck Chemical LLC Atazanavir S4662-50mg
SDP
Supplier:  Selleck Chemical LLC S466250MG
Encompass_Preferred
Atazanavir (Latazanavir Zrivada Reyataz BMS-232632) is an azapeptide and HIV-protease inhibitor that is used in the treatment of HIV infections and AIDS in combination with other anti-HIV agents Atazanavir is a substrate and inhibitor of cytochrome P450 isozyme 3A (CYP3A4) and an inhibitor and inducer of P-glycoprotein
Catalog No. 50-311-0845