Learn More
Medchemexpress LLC Atazanavir sulfate | 229975-97-7 | 99.7% | 802.93 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY173675MG
Encompass_Preferred
Atazanavir (BMS-232632) sulfate is a highly selective and orally active HIV-1 protease inhibitor with blood-brain barrier permeability. Atazanavir sulfate is a substrate and inhibitor of CYP3A4, and an inhibitor of P-glycoprotein (P-gp). Atazanavir sulfate is also a SARS-CoV 3CLpro inhibitor with an IC50 of 3.49 μM. Atazanavir sulfate inhibits cardiac fibrosis, hyperlipidemia and induces malignant glioma death.
  • Highly selective and orally active HIV-1 protease inhibitor with blood-brain barrier permeability.
  • Functions as a substrate and inhibitor of CYP3A4.
  • Acts as an inhibitor of P-glycoprotein (P-gp).
  • Inhibits SARS-CoV 3CLpro with an IC50 of 3.49 μM.
  • Inhibits cardiac fibrosis.
  • Inhibits hyperlipidemia.
  • Induces malignant glioma death.
Catalog No. 50-003-84225