Learn More
Medchemexpress LLC Atazanavir (sulfate) | 229975-97-7 | 99.7% | 802.93 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY17367A5MG
Encompass_Preferred
Atazanavir sulfate is a highly selective and orally active HIV-1 protease inhibitor with blood-brain barrier permeability. It acts as a substrate and inhibitor of CYP3A4, and an inhibitor of P-glycoprotein (P-gp). It also functions as a SARS-CoV 3CLpro inhibitor with an IC50 of 3.49 μM, and inhibits cardiac fibrosis, hyperlipidemia, and induces malignant glioma death.
  • Highly selective and orally active HIV-1 protease inhibitor
  • Blood-brain barrier permeability
  • Substrate and inhibitor of CYP3A4
  • Inhibitor of P-glycoprotein (P-gp)
  • SARS-CoV 3CLpro inhibitor with an IC50 of 3.49 μM
  • Inhibits cardiac fibrosis and hyperlipidemia
  • Induces malignant glioma death
Catalog No. 50-003-84226