Learn More
TARGETMOL CHEMICALS INC ATB107 HYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T10394L5MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ATB107 hydrochloride is a novel and potent inhibitor of indole-3-glycerol phosphate synthase (IGPS) with a KD of 3 uM.
Catalog No. 50-258-1727