Learn More
ATCC ATCC Bordetella pertussis Strain C352 (BAA-3222), Frozen culture, 0.5 mL
Supplier:  ATCC BAA3222
Encompass
Bordetella pertussis strain C352 is a clinical isolate supplied by ATCC (BAA-3222) as a frozen culture for research and strain characterization. The depositor reports mixed resistance to macrolide antibiotics. This material is intended for microbiology research and antimicrobial studies and must be handled under appropriate biosafety conditions.
  • Strain designation C352.
  • Isolated from a human (United States) in 1999.
  • Deposited by CDC Pertussis and Diphtheria Laboratory.
  • Mixed resistance to macrolide antibiotics.
  • Supplied as a frozen culture, 0.5 mL.
  • Intended for research use; follow biosafety procedures.
Catalog No. NC3871998