Learn More
ATCC ATCC CHLA-05-ATRT (CRL-3037) Human atypical teratoid rhabdoid tumor cell line
Supplier:  ATCC CRL3037
Encompass
CHLA-05-ATRT is a human cell line derived from an atypical teratoid rhabdoid tumor of the frontal lobe. It is suited for studies of tumor biology, metabolism, and therapeutic screening, and is compatible with 3D culture and neuroscience models. The line exhibits mixed growth (suspension and aggregates) and shows loss of INI-1 expression by immunohistochemistry.
  • Derived from pediatric brain tumor tissue for disease-relevant research.
  • Exhibits mixed growth as suspension and aggregate cultures.
  • Suitable for metabolic studies, drug screening, and 3D/neuroscience models.
  • Recommended culture: DMEM:F12 with B-27, EGF, and FGF; incubate at 37°C with 5% CO2.
  • Biosafety level 1; male donor origin.
Catalog No. NC3814812