Learn More
ATCC ATCC Hybridoma L243 (HB-55) - Mouse hybridoma producing anti-HLA-DR monoclonal antibody
Supplier:  ATCC HB55L243
Encompass
ATCC Hybridoma L243 (HB-55) is a mouse-derived hybridoma cell line that secretes a monoclonal antibody recognizing HLA-DR. It is provided for research applications that require production of conditioned medium containing the antibody or direct use of secreted antibody in immunoassays and functional studies.
  • Produces monoclonal antibody specific for HLA-DR.
  • Grows in suspension culture.
  • Suitable for generating conditioned medium for antibody purification.
  • Used in flow cytometry, immunostaining, and antigen-blocking assays.
  • Derived from mouse myeloma fusion for stable antibody secretion.
Catalog No. NC3918601