Learn More
ATCC ATCC LAMA84-r (CRL-3348) human CML suspension cell line, imatinib-resistant - 8.0x10^6-1.0x10^7 cells/vial
Supplier:  ATCC CRL3348
Encompass
LAMA84-r (ATCC CRL-3348) is a human chronic myeloid leukemia (CML)-derived suspension cell line resistant to imatinib. Paired with an imatinib-sensitive clone, it is used to study sensitivity and resistance to BCR-ABL-targeted therapies.
  • Human hematopoietic suspension cell line derived from CML in blast crisis.
  • Resistant to imatinib mesylate; paired with an imatinib-sensitive clone for comparison.
  • Approximately 8.0 x 10^6 to 1.0 x 10^7 cells per vial; vial volume 1.0 mL.
  • Viability ≥ 50% on arrival; store frozen in liquid nitrogen vapor.
  • Maintain cultures at 3 x 10^5 to 1 x 10^6 viable cells/mL; renew medium every 2-3 days.
  • Recommended base medium: RPMI-1640 supplemented with fetal bovine serum, L-glutamine, and 1 μM imatinib working stock.
Catalog No. NC3719697