Learn More
ATCC ATCC MicroQuant MQ-60 Microbial Examination of Nonsterile Products Panel (Burkholderia cepacia complex)
Supplier:  ATCC MQ60
Encompass
The MicroQuant MQ-60 Microbial Examination of Nonsterile Products panel is a quantified multi-organism control kit for quality control testing of water-based pharmaceutical products that targets the Burkholderia cepacia complex. It provides single-use cryopreserved pellets and rehydration buffer for streamlined assay setup and reproducible results.
  • Provides five low-CFU challenge organisms relevant to USP testing.
  • Each vial contains 100-1,000 CFU (approximately 10-100 CFU per 100 μL).
  • Each vial supports up to 10 assays.
  • Pack format includes cryopreserved pellet vials and rehydration buffer for immediate rehydration and use.
  • Store at 2 °C to 8 °C; 12-month shelf life when stored properly.
  • Intended for laboratory quality control and nonclinical testing; not for therapeutic, diagnostic, or consumptive use.
Catalog No. NC3853625