Learn More
ATCC ATCC MicroQuant MQ-62 Microbial Examination of Nonsterile Products Panel (Tests for Specified Microorganisms)
Supplier:  ATCC MQ62
Encompass
MicroQuant MQ-62 is a ready-to-use panel designed for the microbial examination of nonsterile products and for performing tests for specified microorganisms. The panel provides precisely quantitated, single-use microbial reference materials to streamline quality control workflows and support USP <62> testing.
  • Single-use, rapidly rehydrating pelleted format.
  • Fridge-stable storage at 2-8 °C for months.
  • Available in high- and low-titer formats for accurate CFU control.
  • Rehydrates and is ready to use in approximately 1 minute.
  • Usable up to 8 hours after rehydration when stored at 2-8 °C.
  • Created from authenticated, passage-zero strains to support regulatory methods.
Catalog No. NC3853621