Learn More
ATCC ATCC Simian Rotavirus Strain SA-11 (VR-1565) - Frozen virus culture, BSL-2 research material
Supplier:  ATCC VR1565
Encompass
ATCC Simian rotavirus (VR-1565), strain SA-11, is a frozen Rotavirus A culture supplied for laboratory research. The strain is serotype 3 (G3) for VP7, derived from a prior isolate after mycoplasma removal, and is propagated in MA-104 Clone 1 cells (embryonic rhesus monkey kidney cells). Store at -70°C or colder. Intended for research use only; handle using appropriate BSL-2 precautions.
  • Simian rotavirus strain SA-11.
  • Serotype 3 (G3) for VP7.
  • Frozen format for long-term storage.
  • Store at -70°C or colder.
  • Nonpathogenic for mice.
  • Handle under biosafety level 2 conditions.
  • Propagate in MA-104 cells using serum-free medium with trypsin.
Catalog No. NC3800424