Learn More
ATCC ThawReady™ THP-1
Supplier:  ATCC TIB202AR
Encompass
ATCC ThawReady™ THP-1 monocytes can be used at thaw, eliminating lengthy cell expansion. These assay ready cells allow you to acquire your data in hours rather than weeks, eliminating the need for cell expansion or banking.
Catalog No. NC3456095