Learn More
ATCC ThawReady™ THP-1 NF-kB-Luc2; 10 pack
Supplier:  ATCC TIB202NFLUC2AR10
Encompass
ATCC ThawReady™ THP-1 NF-kB-Luc2 monocytes can be used at thaw, eliminating lengthy cell expansion. These assay ready cells allow you to acquire your data in hours rather than weeks, eliminating the need for cell expansion or banking. Additionally, these cells express luciferase under the control of the NF-kB reporter, allowing sensitive and quantitative quantification of NF-kB activation.
Catalog No. NC3456090
Only null left
Add to Cart
Request a Quote