Learn More
ATCC ThawReady™ THP-1 NF-kB-Luc2
Supplier:  ATCC TIB202NFKBLUC2AR
Encompass
ATCC ThawReady™ THP-1 NF-kB-Luc2 monocytes can be used at thaw, eliminating lengthy cell expansion. These assay ready cells allow you to acquire your data in hours rather than weeks, eliminating the need for cell expansion or banking. Additionally, these cells express luciferase under the control of the NF-kB reporter, allowing sensitive and quantitative quantification of NF-kB activation.
Catalog No. NC3456093