Learn More
INTACT GENOMICS INC ATCC15834 Agrobacterium Chemically Competent Cells 18x50µl
Supplier:  INTACT GENOMICS INC 107518
Encompass
Intact Genomics (ig) ATCC15834 Agrobacterium Chemically Competent Cells are made from a specific strain of Rhizobium rhizogenes (formerly Agrobacterium rhizogenes), Agrobacterium rhizogenes pRi15834 (agropine type). Agrobacterium rhizogenes is a soil-borne gram-negative bacterium that can infect most dicotyledons, a few monocotyledons and some gymnosperms. ATCC15834 Chemically Competent Agrobacterium are optimized for the highest transformation efficiencies and are useful for transgenic operations of legumes, tobacco, variety of grasses and other plants. ATCC15834 Agrobacterium rhizogenes strain contains pRi15834 agrobacterium-type Ri plasmid and displays resistance to Rifampicin.Product Includes and Storage:ATCC15834 Agrobacterium Chemically Competent Cells: -80 ºCpCAMBIA1391z control DNA: -20 ºCAgro Recovery medium: 4 ºC
Catalog No. 50-000-82886