Learn More
Cayman Chemical Atenolol 10mg
SDP
Supplier:  Cayman Chemical 1862310 MG
Encompass_Preferred
An enantiomer of (±)-atenolol; inhibits radioligand binding to β-ARs on sarcolemma-enriched membranes (Ki = 8.61 µM); has no effect on blood pressure in spontaneously hypertensive rats
Catalog No. 50-298-0128