Learn More
Cayman Chemical ATG4B MoncLNal ANTIBDYCl 100ug
SDP
Supplier:  Cayman Chemical 25963100 UG
Encompass_Preferred
Immunogen: Full-length human recombinant ATG4B protein • Host: Mouse • Species Reactivity: Human • Cross Reactivity: (-)-ATG8 • Applications: ELISA, IHC, IP, and WB • MW = ~44 kDa
Catalog No. 50-293-5631