Learn More
Selleck Chemical LLC Atglistatin 10mg 1469924-27-3
SDP
Supplier:  Selleck Chemical LLC S736410MG
Encompass_Preferred
Atglistatin is a highly potent, and selective inhibitor of adipose triglyceride lipase (ATGL) with IC50 of 0.7 ?M, high selectivity over other key metabolic lipases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4693