Learn More
Air Techniques Intl PAO-4 Reagent, 1 gallon
SDP
Supplier:  Air Techniques Intl T0001075
Encompass_Preferred
This reagent offers a U.S. Food & Drug Administration-approved alternative to DOP in FDA-regulated filter leakage testing applications. PAO is approved by the U.S. Army Surgeon General and is used by the United States military as a substitute for DOP in their filter and mask testing processes.
Catalog No. NC0022435