Learn More
Medchemexpress LLC Aticaprant | 1174130-61-0 | MFCD22572355 | 99.6% | 418.5 g/mol | C26H27FN2O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101718100MG
Encompass_Preferred
Aticaprant is a selective, centrally penetrant kappa-opioid receptor (KOR) antagonist supplied for research use. It is used in pharmacology studies to investigate KOR-mediated pathways and has reported high affinity for the KOR.
  • Potent kappa-opioid receptor antagonist with Ki ≈ 0.807 nM.
  • Selective for kappa over mu and delta opioid receptors.
  • High purity research-grade powder (≈99.6%).
  • Suitable for in vitro and in vivo studies; intended for research use only.
  • Supplied as a 100 mg solid pack with cold-storage recommendations.
  • Molecular weight 418.5 g/mol; formula C26H27FN2O2.
Catalog No. 50-002-47614