Learn More
Medchemexpress LLC HY-101718 50mg , Aticaprant CAS:1174130-61-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10171850MG
Encompass_Preferred
HY-101718 50mg Aticaprant CAS: 1174130-61-0 Aticaprant (CERC-501) is a potent and centrally-penetrant kappa opioid receptor antagonist with a Ki of 0.807 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9292