Learn More
TARGETMOL CHEMICALS INC Atipamezole 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T676625MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Atipamezole (Antisedan) is a synthetic alpha2 adrenergic receptor antagonist. It has also been researched in humans as a potential anti-Parkinsonian drug. Purity 99.76%
Catalog No. 50-287-6361