Learn More
ABclonal Technology ATL3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A1219620UL
Encompass_Preferred
This gene encodes a member of a family of dynamin-like, integral membrane GTPases. The encoded protein is required for the proper formation of the network of interconnected tubules of the endoplasmic reticulum. Mutations in this gene may be associated with hereditary sensory neuropathy type IF. Alternatively spliced transcript variants that encode distinct isoforms have been described.
Catalog No. 50-001-13832