Learn More
Waters Corp Waters Atlantis dC18 Column, 100Å, 3 μm, 3 mm X 50 mm
Supplier:  Waters Corp 186001389
Encompass_Preferred
The Atlantis dC18 Column is a universal, silica-based, reversed-phase C18 column for polar compound retention. It offers balanced retention for polar and non-polar substances, optimized for highly aqueous mobile phases.
  • Di-functionally bonded C18 ligands for highly aqueous mobile phases (up to 100% water).
  • Greater retention of polar compounds without excessive hydrophobic retention.
  • Universal C18 column, fully LC/MS compatible.
  • Improved pH stability and water mobile phase compatibility.
  • Separates polar chemicals and supports reversed-phase applications.
  • Low pH stability and extended column lifetime from difunctional silane bonding.
  • Superior peak shape and dewetting resistance for reversed-phase HPLC.
  • Operates 3–7 pH range and up to 6000 psi (415 Bar) pressure.
Catalog No. 50-819-127