Learn More
Waters Corp Column, dC18, 100 Å, 5 μm, 4.6 mm × 250 mm, eConnect
Supplier:  Waters Corp 186001346RF
Encompass_Preferred
This column is equipped with an electronic eConnect column tag that contains unique, column-specific information. These NFC-enabled device-tagged columns are designed to automatically identify, track, and digitally trace column use history. While eConnect columns can be used on any HPLC, their full advantages are realized when used with the Waters Alliance iS System running Empower software.
  • Equipped with an electronic eConnect column tag for unique column-specific information.
  • NFC-enabled device-tagged columns for automatic identification, tracking, and digital tracing of column use history.
  • Compatible with any HPLC, with enhanced benefits when used with the Alliance iS System running Empower software.
Catalog No. 50-239-1874