Learn More
Waters Corp Waters Atlantis dC18 Column, 3 μm, 3 mm X 100 mm, 100 Å
Supplier:  Waters Corp 186001303
Encompass_Preferred
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns, featuring di-functionally bonded C18 ligands optimized for highly aqueous mobile phases, including 100% water. This analytical column balances retention for polar and nonpolar chemicals while minimizing hydrophobic compound retention, offering improved pH stability.
  • Universal, silica-based, reversed-phase C18 column for polar compound retention.
  • Optimized for highly aqueous mobile phases, including 100% water, with di-functionally bonded C18 ligands.
  • Compatible with both LC and MS.
  • Balances retention for polar and nonpolar chemicals while minimizing hydrophobic compound retention.
  • Improved pH stability and longer column lifetime due to di-functional silane bonding chemistry.
  • Superior peak form and improved polar compound retention.
Catalog No. 50-780-650