Learn More
Waters Corp Waters Corporation Atlantis dC18 OBD Column, 5 μm, 19 x 100 mm
Supplier:  Waters Corp 186001367
Encompass_Preferred
The Atlantis dC18 OBD Column is a preparative column designed for chromatography applications. It utilizes Optimum Bed Density (OBD) technology to provide predictable, uniform density profiles and eliminate voids during the final capping process.
  • Exceptional resistance to mechanical chromatographic bed failure.
  • Delivers consistent column-to-column performance.
  • Extends column lifetimes and reduces costs.
  • Allows for direct scaling from UPLC,™ UHPLC, or HPLC screening to lab-scale purification.
  • Ensures chromatographic equivalence between analytical and preparative dimensions for successful method scaling.
Catalog No. 50-780-675