Learn More
Waters Corp Atlantis dC18 Column, 100Å, 5 µm, 3 mm X 100 mm, 1/pk
Supplier:  Waters Corp 186001305
Encompass_Preferred
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns that are used for polar compound retention. The Atlantis dC18 columns feature di-functionally bonded C18 ligands that have been optimized for use with highly aqueous mobile phases, including 100% water.
Catalog No. 50-780-651