Learn More
Waters Corp Atlantis Method Validation Kit, BEH C18 AX, 1.7 μm, 2.1 × 150 mm
Supplier:  Waters Corp 186009372
Encompass_Preferred
The method validation kit contains three columns, each packed with a different lot of material. The Atlantis Premier BEH C18 AX columns offer excellent retention for polar acidic analytes under reversed-phase conditions. The reversed-phase/anion-exchange mixed-mode chemistry is stable from pH 2-10 and provides complementary selectivity to traditional C18 stationary phases, which is beneficial for method development.
  • Contains three columns, each packed with a different lot of material.
  • Provides excellent retention for polar acidic analytes under reversed-phase conditions.
  • Mixed-mode chemistry is stable from pH 2-10.
  • Offers complementary selectivity to traditional C18 stationary phases.
  • Beneficial for method development.
Catalog No. 50-239-2931