Learn More
Waters Corp HPLC Column, BEH C18 AX, 2.5 μm, 2.1 x 100 mm, VanGuard FIT
Supplier:  Waters Corp 186009378
Encompass_Preferred
These columns provide excellent retention for polar acidic analytes under reversed-phase conditions. Their reversed-phase/anion-exchange mixed-mode chemistry is stable from pH 2-10, offering complementary selectivity to traditional C18 stationary phases. The VanGuard FIT Column format is an all-in-one, guard-column design engineered to reduce dead volume and increase column lifetime.
  • Separates polar acidic analytes via HPLC.
  • Anion-exchange chemistry supports analytes from 2 to 10 pH.
  • VanGuard FIT Technology prevents signal loss.
  • Reduces dead volume, increases efficiency.
  • Offers complementary selectivity to C18 phases.
  • Maximizes throughput for reliable data.
  • Effective for amphiphilic compound separations.
  • Stable BEH particle morphology resists acids and bases.
  • Compatible with 100% aqueous mobile phases.
Catalog No. 50-239-1475