Learn More
Waters Corp Atlantis VanGuard FIT Cartridge, Mixed-mode, 5 μm, 3.9 x 5 mm
Supplier:  Waters Corp 186009422
Encompass_Preferred
These mixed-mode reversed-phase/anion-exchange guard cartridges are designed to protect the analytical column from matrix and sample contaminants. They prevent chemical contaminants and suspended particulates from entering the analytical column, extending its lifespan and dependability.
  • Mixed-mode chemistry offers high retention for polar acidic analytes in reversed-phase conditions.
  • Provides complementary selectivity to conventional C18 stationary phases.
  • VanGuard FIT (Fast Installation Technology) cartridge design reduces dead volume and extends column longevity.
  • Allows for easy and quick installation, improving workflow efficiency.
  • Suitable for both analytical and preparative applications.
  • Designed for use with both polar and nonpolar compounds due to balanced hydrophobicity and strong cation exchange functionality.
Catalog No. 50-239-2625