Learn More
Waters Corp VanGuard FIT Column, Atlantis Premier BEH C18 AX, 5 μm, 4.6 x 150 mm
Supplier:  Waters Corp 186009412
Encompass_Preferred
The Atlantis Premier BEH C18 AX VanGuard FIT Column offers excellent retention for polar acidic analytes using stable reversed-phase/anion-exchange mixed-mode chemistry, beneficial for method development. Its guard-column design reduces dead volume and extends analytical column lifetime by preventing contaminants, improving analytical performance.
  • Excellent retention for polar acidic analytes in reversed-phase.
  • Stable reversed-phase/anion-exchange mixed-mode chemistry (pH 2–10).
  • Complementary selectivity for method development.
  • All-in-one guard-column design; reduces dead volume.
  • Extends analytical column lifetime, prevents contaminants.
  • Readily replaceable to restore separation performance.
  • Filters chemical and particle pollutants from mobile phase.
  • Suitable for biological fluid cleanup or temperature control.
  • Connects directly to an inlet.
Catalog No. 50-239-2996