Learn More
Waters Corp Column, Premier BEH C18 AX VanGuard FIT, 2.5 μm, 2.1 x 150 mm
Supplier:  Waters Corp 186009379
Encompass_Preferred
The Atlantis Premier BEH C18 AX Columns offer excellent retention for polar acidic analytes under reversed phase conditions. Their reversed phase/anion-exchange mixed-mode chemistry is stable from pH 2-10 and provides complementary selectivity, beneficial for methods development. The VanGuard FIT Column format is an all-in-one guard-column design that reduces dead volume and extends column lifetime.
  • Excellent retention for polar acidic analytes.
  • Mixed-mode chemistry (pH 2-10) provides complementary selectivity.
  • Guard-column design reduces dead volume and extends column life.
  • Prevents unwanted sample matrix and particle introduction.
  • Extends analytical column operational lifetime.
  • High-performance packing for biological fluid cleanup, temperature control.
  • Maximum operating limits: 10 pH, 60 °C, 18,000 psi (1240 Bar).
Catalog No. 50-239-1717