Learn More
Waters Corp Waters Atlantis Premier BEH Z-HILIC Column, 1.7 μm, 2.1 mm x 50 mm, Sulfobetaine
Supplier:  Waters Corp 186009978
Encompass_Preferred
Atlantis Premier BEH Z-HILIC Columns enhance polar analyte retention and offer distinct selectivity for HILIC methods development. Featuring rugged ethylene bridged hybrid particles, they provide pH stability from 2–10 and utilize MaxPeak High Performance Surface hardware to minimize unwanted analyte/surface interactions.
  • Increase polar analyte retention and offer distinct HILIC selectivity.
  • Built with rugged ethylene bridged hybrid particles, stable from pH 2–10.
  • MaxPeak High Performance Surface reduces analyte/surface interactions for improved peak shape and signal.
  • Chromatographically tested batches ensure reproducible performance.
  • eCord Intelligent Chip stores column performance history and usage data.
Catalog No. NC1994237