Learn More
Waters Corp Waters BEH Z-HILIC Column, 5 μm, 4.6 mm x 150 mm
Supplier:  Waters Corp 186010008
Encompass_Preferred
Atlantis Premier BEH Z-HILIC Columns enhance polar analyte retention and offer distinct selectivity for HILIC methods development. Constructed with durable ethylene bridged hybrid particles, these columns provide pH stability from pH 2 to 10. MaxPeak High Performance Surface hardware technology significantly reduces unwanted interactions between analytes and the column surface, which can cause poor peak shape and decreased signal intensity.
  • Increased polar analyte retention
  • Distinct selectivity compared to other HILIC chemistries
  • Built using rugged ethylene bridged hybrid particles
  • pH stable from pH 2 to 10
  • MaxPeak High Performance Surface hardware reduces unwanted analyte interactions, mitigating poor peak shape and preventing signal loss
Catalog No. 50-222-4410