Learn More
Waters Corp Waters Atlantis Prep HILIC Column, 5 μm, 19 x 100 mm
Supplier:  Waters Corp 186003958
Encompass_Preferred
Waters Atlantis Prep HILIC Column is an OBD Column designed for HILIC separations. It features a 5 μm particle size and dimensions of 19 x 100 mm.
  • Excellent retention for very polar, basic, water soluble analytes.
  • Designed and tested for HILIC separations using mobile phases containing high concentration of organic solvent.
Catalog No. 50-782-088