Learn More
Waters Corp Atlantis T3 Column, 100Å, 3 μm, 4.6 mm x 150 mm
Supplier:  Waters Corp 186003729RF
Encompass_Preferred
This Atlantis T3 HPLC column incorporates eConnect technology, featuring a permanently attached electronic tag for unique product identification and convenient information access via NFC. It aids in streamlining laboratory workflows through automated data entry and usage tracing with compatible systems.
  • Uses a permanent electronic tag for product identification.
  • Stores product-specific data, including serial numbers and descriptions.
  • NFC technology allows easy access to information.
  • Tag data is automatically imported and searchable by compatible software.
  • Simplifies setup and reduces manual data entry; ready for use out of the box.
  • Tamper-proof design enhances compliance and system setup.
  • Strategic tag placement ensures reliable detection and prevents temperature interference.
  • Equivalent to untagged columns, guaranteeing identical quality and performance.
Catalog No. 50-239-2512