Learn More
Waters Corp Waters Atlantis T3 Column, C18, 3 μm, 2.1 mm x 100 mm
Supplier:  Waters Corp 186003718
Encompass_Preferred
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns for balanced retention of polar and hydrophobic molecules. They are ideal for developing separations of both polar and non-polar compounds, offering superior retention, excellent peak shape, and exceptional chemical stability across a wide pH range.
  • Provides balanced retention for polar and hydrophobic molecules.
  • Offers superior retention for polar compounds through optimized pore diameter, C18 ligand density, and proprietary end-capping.
  • Maintains industry-leading peak shape and efficiency for all analytes.
  • Exhibits exceptional chemical stability across a wide pH range.
  • Ensures extended column lifetime at low pH due to resistance to ligand cleavage.
  • Manufactured in a cGMP, ISO 9001:200 certified plant; each column individually tested.
Catalog No. 50-781-981