Learn More
Waters Corp Atlantis T3 Column, 100Å, 3 µm, 2.1 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186003719
Encompass_Preferred
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns that provide balanced retention of polar and hydrophobic molecules and should be your first choice when developing separations of polar and non-polar compounds.
Catalog No. 50-781-982