Learn More
Waters Corp Waters Atlantis T3 Column, 100 Å, 3 μm, 3 mm x 100 mm
Supplier:  Waters Corp 186003722
Encompass_Preferred
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns for balanced retention of polar and hydrophobic molecules, ideal for developing separations of polar and non-polar compounds.
  • T3 bonding technology promotes polar compound retention and mobile phase compatibility with a trifunctional C18 alkyl phase.
  • Efficient end-capping is superior to traditional trimethyl silane (TMS) end-capping.
  • Superior performance offers enhanced column performance, improved lifetime, peak shape, and stability.
  • HPLC methods can be easily transferred to UPLC methods due to T3 bonding.
  • Extended column lifetimes under low pH conditions; T3 bonding protects the silica substrate from dissolution.
Catalog No. 50-781-985