Learn More
Waters Corp Atlantis T3 Column, 3 μm Particle, 100 Å Pore, 4.6 mm ID, 75 mm Length
Supplier:  Waters Corp 186003727
Encompass_Preferred
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns designed to provide balanced retention of polar and hydrophobic molecules. They are recommended for developing separations of polar and non-polar compounds.
  • T3 bonding method for polar chemical retention and aqueous compatibility
  • Patented T3 end-capping for enhanced efficiency over conventional TMS end-capping
  • Improved column longevity, peak shape, and stability
  • Extended life at low pH and better peak shape for bases at pH 7
  • Performs well over a wide pH range (2 to 8 pH)
Catalog No. 50-781-990