Learn More
Waters Corp Waters Column, 100Å, 3 μm, 4.6 mm x 100 mm
Supplier:  Waters Corp 186003728
Encompass_Preferred
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns that provide balanced retention of polar and hydrophobic molecules. They are recommended as a first choice for developing separations of polar and non-polar compounds. The column offers superior performance across a wide pH range and is designed for longer lifetimes at low pH and provides improved peak shape for amine-containing bases at pH 7.
  • Advanced T3 bonding process promotes polar compound retention and aqueous mobile phase compatibility.
  • Proprietary T3 end-capping is more effective than traditional trimethyl silane (TMS) end-capping.
  • Unique end-capping and T3 bonding enhance column performance, lifetime, peak shape, and stability.
  • Excellent alternative to traditional C18 reversed-phase LC columns, offering good retention and selectivity for neutral and hydrophobic compounds, in addition to polar analytes.
Catalog No. 50-781-991