Learn More
Waters Corp Atlantis T3 Column, 5 μm, 4.6 mm x 75 mm, 100 Å
Supplier:  Waters Corp 186003745
Encompass_Preferred
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns. They offer balanced retention for both polar and hydrophobic molecules, making them ideal for separating polar and non-polar compounds. Advanced T3 bonding technology, utilizing a trifunctional C18 alkyl phase, enhances polar compound retention and mobile phase compatibility.
  • Designed for balanced retention of polar and hydrophobic molecules.
  • Features advanced T3 bonding technology with a trifunctional C18 alkyl phase.
  • Promotes enhanced polar compound retention and mobile phase compatibility.
  • Utilizes efficient T3 end-capping for improved column performance and lifetime.
  • Ensures superior peak shape and stability, especially under low pH conditions.
  • Protects the silica substrate from dissolution, extending column lifespan.
  • Simplifies the conversion of HPLC methods to UPLC techniques.
Catalog No. 50-782-008