Learn More
AGI NORTH AMERICA INC Syrris Atlas Syringe Pump for precise dosing, flow rates from 5μL to 200mL/min - ideal for autosampling, pH control and full data logging capabilities
SDP
Supplier:  AGI NORTH AMERICA INC 10003ARBF
Encompass
Atlas XL Syringe Pump offers a small footprint and is lightweight to be easily moved between lab benches.Functions -Dual-Dosing Mode allows the use of each syringe to dose independently-Continuous-Dosing Mode combines both dosing channels to provide a single continuous flow-Autosampling Mode allows the collection of 6 samples from the process at user defined times-pH-controlled dosing automatically doses acid or base to keep the pH of the reaction within a user defined range-Multi-port valves allow the dosing of multiple reagents -All data is recorded (dosed amounts, sensor readings) for easy analysisIncluded: -25mL syringes as standard (syringe sizes from 50μL to 5mL are available)-4 port valves (ports are channels which can be set as inlets or outlets)-Starter kit including tubing and blanking plugsOrder informationTo perform autosampling, pH control and temperature-dependent dosing additional Syrris sensors, nodes and modules are required.
Catalog No. NC2417590
Only null left
Add to Cart
Request a Quote